Oxalic acid 5% (w/v) in water

Supplier: BeanTown Chemical
Danger

144470-1L 144470-4L
BT144470-1LEA 46.54 USD
BT144470-1L BT144470-4L
Oxalic acid 5% (w/v) in water
Oxalic acid

CAS: 144-62-7; EC No: 205-634-3; MDL No: MFCD00002573 UN No: UN3265; Haz Class: 8; Packing Group: III Liquid; Linear Formula: HOOCCOOH; Molecular Formula: C2H2O4; MW: 90.04


  • Liquid

Formula: C₂H₂O₄
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR