Tin(II) 2-ethylhexanoate, Technical Grade

Supplier: BeanTown Chemical

Synonyms: 2-Ethylcaproic acid tin (II) salt, Tin bis(2-ethylhexanoate), Stannous 2-ethylhexanoate, 2-Ethylhexanoic acid tin (II) salt

144265-100G 144265-2KG 144265-500G
BT144265-100GEA 34.76 USD
BT144265-100G BT144265-2KG BT144265-500G
Tin(II) 2-ethylhexanoate, Technical Grade
Tin (II) 2-ethylhexanoate

CAS: 301-10-0; EC No: 206-108-6; MDL No: MFCD00002676; RTECS: MO7870000 Liquid; Molecular Formula: C16H30O4Sn; MW: 405.11 Melting Point: 9°; Flash point: 110°C (230°F) Density (g/mL): 1.251; Refractive Index: 1.494


  • RTECS: MO7870000
  • Refractive index: 1.494
  • Liquid

Formula: C₁₆H₃₀O₄Sn
Melting Pt: 9 °C
Density: 1.251 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00002676
CAS Number: 301-10-0
EINECS: 206-108-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR