Phenol red

Supplier: BeanTown Chemical

Synonyms: 3,3-Bis(4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide

144260-10G 144260-250G 144260-50G
BT144260-10GEA 30 USD
BT144260-10G BT144260-250G BT144260-50G
Phenol red
Phenol red

CAS: 143-74-8; EC No: 205-609-7; MDL No: MFCD00003552; RTECS: SJ7490000 Powder; Molecular Formula: C19H14O5S; MW: 354.38 Melting Point: 300°


  • RTECS: SJ7490000
  • Powder

Formula: C₁₉H₁₄O₅S
Melting Pt: >300 °C
MDL Number: MFCD00003552
CAS Number: 143-74-8
EINECS: 205-609-7
UN: 1671
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR