Tin (II) trifluoromethanesulfonate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Tin trifluoromethanesulfonate, Tin(II) trifluoromethanesulfonate, Tin trifluoromethanesulphonate, Tin triflate, Tin(II) trifluoromethanesulphonate
144150-1G
144150-5G
BT144150-1GEA
57.08
USD
BT144150-1G
BT144150-5G
Tin (II) trifluoromethanesulfonate ≥97%
Tin (II) trifluoromethanesulphonate
CAS: 62086-04-8; MDL No: MFCD00191251
UN No: UN3288; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: (CF3SO3)2Sn; Molecular Formula: C2F6O6S2Sn; MW: 416.85
Melting Point:
- Moisture Sensitive
- Powder
Formula:
C₂F₆O₆S₂Sn Melting Pt: >300 °C |
MDL Number:
MFCD00191251 CAS Number: 62086-04-8 UN: 3288 ADR: 6.1,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...