L-Methionine ≥98%
Supplier: BeanTown Chemical
Synonyms:
(S)-(-)-2-Amino-4-(methylthio)butyric acid, (S)-(-)-Methionine, L(-)-Methionine, (S)-(-)-2-Amino-4-(methylthio)butanoic acid, H-Met-OH, Methionine, (S)-2-Amino-4-(methylthio)butanoic acid
144115-100G
144115-2.5KG
144115-500G
BT144115-100GEA
65.79
USD
BT144115-100G
BT144115-2.5KG
BT144115-500G
L-Methionine ≥98%
L-Methionine
CAS: 63-68-3; EC No: 200-562-9; MDL No: MFCD00063097; RTECS: PD0457000
Powder; Molecular Formula: C5H11NO2S; MW: 149.21
Melting Point: 284° (decomposes)
Optical Rotation: [α]20/D +23.7±0.5°, c = 5% in 5M HCl
- RTECS: PD0457000
- Optical rotation: [α]20/D +23.7±0.5°, c = 5% in 5M HCl
- Powder
Formula:
C₅H₁₁NO₂S MW: 149.21 g/mol Boiling Pt: 278 Melting Pt: 136 (dec.) |
MDL Number:
MFCD00063097 CAS Number: 63-68-3 EINECS: 200-562-9 Merck Index: 13,06004 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...