6-Nitro-o-toluidine ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Nitro-2-methylaniline, 2-Amino-3-nitrotoluene, 2-Methyl-6-nitroaniline
144095-100G
144095-25G
BT144095-100GEA
313
USD
BT144095-100G
BT144095-25G
6-Nitro-o-toluidine ≥98%
6-Nitro-o-toluidine
CAS: 570-24-1; EC No: 209-329-6; MDL No: MFCD00007744
UN No: UN2660; Haz Class: 6.1; Packing Group: III
Crystalline; Molecular Formula: C7H8N2O2; MW: 152.15
Melting Point: 93-96°
- Crystalline
Formula:
C₇H₈N₂O₂ Melting Pt: 93…96 °C |
MDL Number:
MFCD00007744 CAS Number: 570-24-1 EINECS: 209-329-6 UN: 2660 ADR: 6.1,III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...