Potassium dihydrogen phosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: Monopotassiumphosphate, Potassium orthophosphate monobasic, prim.-Potassium phosphate, Potassium dihydrogenorthophosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate

144090-10KG 144090-2.5KG 144090-500G
BT144090-10KGEA 339 USD
BT144090-10KG BT144090-2.5KG BT144090-500G
Potassium dihydrogen phosphate ≥98%
Potassium dihydrogen phosphate

CAS: 7778-77-0; EC No: 231-913-4; MDL No: MFCD00011401; RTECS: TC6615500 Crystalline; Linear Formula: KH2PO4; MW: 136.09 Melting Point: 253° Density (g/mL): 2.338 Hygroscopic


  • RTECS: TC6615500
  • Hygroscopic
  • Crystalline

Formula: KH₂PO₄
MW: 136.09 g/mol
Boiling Pt: 100
Melting Pt: 253
Density: 2.338 g/mL
MDL Number: MFCD00011401
CAS Number: 7778-77-0
EINECS: 231-913-4
Merck Index: 13,07744

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR