Palladium(II) iodide ≥99.9%
Supplier: BeanTown Chemical
144075-1G
144075-5G
BT144075-1GEA
79.18
USD
BT144075-1G
BT144075-5G
Palladium(II) iodide ≥99.9%
Palladium (II) iodide
CAS: 7790-38-7; EC No: 232-203-7; MDL No: MFCD00011171
Powder; Linear Formula: PdI2; MW: 360.23
Melting Point: 350° (decomposes)
Density (g/mL): 6.003
- Powder
Formula:
I₂Pd Melting Pt: 350 °C (dec.) Density: 6.003 g/mL |
MDL Number:
MFCD00011171 CAS Number: 7790-38-7 EINECS: 232-203-7 |
Specification Test Results
Assay | 99.9% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...