2-Amino-6-chloro-3-nitropyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine

144010-100G 144010-25G
BT144010-100GEA 234 USD
BT144010-100G BT144010-25G
2-Amino-6-chloro-3-nitropyridine ≥98%
2-Amino-6-chloro-3-nitropyridine

CAS: 27048-04-0; EC No: 248-188-5; MDL No: MFCD00044350 UN No: UN2811; Haz Class: 6.1; Packing Group: III Powder; Molecular Formula: C5H4ClN3O2 ; MW: 173.56


  • Powder

Formula: C₅H₄ClN₃O₂
MW: 173.56
MDL Number: MFCD00044350
CAS Number: 27048-04-0
EINECS: 248-188-5
ADR: 6.1 III

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR