Ethyl benzoate ≥99%
Supplier: BeanTown Chemical
Synonyms:
Benzoic acid ethyl ester
143935-100G
143935-2.5KG
143935-500G
BT143935-100GEA
28.3
USD
BT143935-100G
BT143935-2.5KG
BT143935-500G
Ethyl benzoate ≥99%
Ethyl benzoate
CAS: 93-89-0; EC No: 202-284-3; MDL No: MFCD00009109; RTECS: DH0200000
Liquid; Linear Formula: C6H5COOCH2CH3; Molecular Formula: C9H10O2; MW: 150.18
Melting Point: -34°; Boiling Point: 212°; Flash point: 93°C (199°F)
Density (g/mL): 1.045; Refractive Index: 1.504
- RTECS: DH0200000
- Refractive index: 1.504
- Liquid
Formula:
C₉H₁₀O₂ Boiling Pt: 212 °C Melting Pt: –34 °C Density: 1.045 g/mL Flash Pt: 93 °C (199 °F) |
MDL Number:
MFCD00009109 CAS Number: 93-89-0 EINECS: 202-284-3 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...