(3-Aminopropyl)trimethoxysilane ≥97%

Supplier: BeanTown Chemical

Synonyms: 3-(Trimethoxysilyl)propylamine, 3-(Trimethoxysilyl)propan-1-amine, Trimethoxy(3-aminopropyl)silane, 3-Aminopropyltrimethoxysilane

143875-100G 143875-500G
BT143875-100GEA 56.3 USD
BT143875-100G BT143875-500G
(3-Aminopropyl)trimethoxysilane ≥97%
(3-Aminopropyl)trimethoxysilane

CAS: 13822-56-5; EC No: 237-511-5; MDL No: MFCD00008206 UN No: UN2735; Haz Class: 8; Packing Group: II Liquid; Molecular Formula: C6H17NO3Si; MW: 179.29 Boiling Point: 91-92°/15 mmHg; Flash point: 90°C (194°F) Density (g/mL): 1.027; Refractive Index: 1.424 Air Sensitive, Moisture Sensitive


  • Refractive index: 1.424
  • Air Sensitive, Moisture Sensitive
  • Liquid

Formula: H₂NCH₂CH₂CH₂Si(OCH₃)₃
MW: 179.29 g/mol
Boiling Pt: 194
Density: 1.027 g/mL
Flash Pt: 90 °C (194 °F)
MDL Number: MFCD00008206
CAS Number: 13822-56-5
EINECS: 237-511-5
ADR: 8 II

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR