Potassium thiocyanate ≥98%
Supplier: BeanTown Chemical
CAS: 333-20-0; EC No: 206-370-1; MDL No: MFCD00011413; RTECS: XL1925000
Crystalline; Linear Formula: KSCN; MW: 97.18
Melting Point: 173°; Boiling Point: 500° (decomposes)
Density (g/mL): 1.89
Light Sensitive, Moisture Sensitive
- RTECS: XL1925000
- Light Sensitive, Moisture Sensitive
- Crystalline
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...