Copper(II) sulfide ≥99.5%

Supplier: BeanTown Chemical

Synonyms: Copper sulfide, Cupric sulfide, Copper sulphide, Cupric sulphide, Copper(II)sulfide, Copper(II)sulphide

143780-1KG 143780-250G 143780-50G
BT143780-1KGEA 626 USD
BT143780-1KG BT143780-250G BT143780-50G
Copper(II) sulfide ≥99.5%
Copper (II) sulphide

CAS: 1317-40-4; EC No: 215-271-2; MDL No: MFCD00016066 -200 Mesh Powder; Molecular Formula: CuS; MW: 95.60 Melting Point: 220° (decomposes) Density (g/mL): 4.6 Air Sensitive, Moisture Sensitive


  • Air Sensitive, Moisture Sensitive
  • -200 Mesh Powder

Formula: CuS
Melting Pt: 220 °C (dec.)
Density: 4.6 g/mL
MDL Number: MFCD00016066
CAS Number: 1317-40-4
EINECS: 215-271-2

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR