Dimethyl phosphonate ≥98%
Supplier: BeanTown Chemical
Synonyms:
Dimethyl phosphite
143775-100ML
143775-500ML
BT143775-100MLEA
55.13
USD
BT143775-100ML
BT143775-500ML
Dimethyl phosphonate ≥98%
Dimethyl phosphonate
CAS: 868-85-9; EC No: 212-783-8; MDL No: MFCD00044633; RTECS: SZ7710000
UN No: UN3278; Haz Class: 6.1; Packing Group: III
Liquid; Molecular Formula: C2H7O3P; MW: 110.05
Boiling Point: 170-171°; Flash point: 70°C (158°F)
Density (g/mL): 1.2; Refractive Index: 1.402
Moisture Sensitive
- RTECS: SZ7710000
- Refractive index: 1.402
- Moisture Sensitive
- Liquid
Formula:
C₂H₇O₃P MW: 110.05 g/mol Boiling Pt: 170…171 °C Density: 1.2 g/mL Flash Pt: 70 °C (158 °F) |
MDL Number:
MFCD00044633 CAS Number: 868-85-9 EINECS: 212-783-8 ADR: 6.1 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...