Boron tribromide ≥99%

Supplier: BeanTown Chemical


ampouled under argon

143760-50G
BT143760-250GEA 511.41 USD
BT143760-250G BT143760-50G
Boron tribromide ≥99%
Boron tribromide

CAS: 10294-33-4; EC No: 233-657-9; MDL No: MFCD00011312; RTECS: ED7400000 UN No: UN2692; Haz Class: 8 (6.1); Packing Group: I Liquid; Molecular Formula: BBr3; MW: 250.52 Melting Point: -46°; Boiling Point: 90° Density (g/mL): 2.6 Moisture Sensitive


  • RTECS: ED7400000
  • Moisture Sensitive
  • Liquid

Formula: BBr₃
MW: 250.52 g/mol
Boiling Pt: 90 °C
Melting Pt: –46 °C
Density: 2.6 g/mL
MDL Number: MFCD00011312
CAS Number: 10294-33-4
EINECS: 233-657-9
UN: 2692
ADR: 8,I

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR