Sodium thiosulfate pentahydrate ≥99%
Supplier: BeanTown Chemical
143715-10KG
143715-2.5KG
143715-500G
BT143715-10KGEA
212
USD
BT143715-10KG
BT143715-2.5KG
BT143715-500G
Sodium thiosulfate pentahydrate ≥99%
Sodium thiosulphate pentahydrate
CAS: 10102-17-7; EC No: 231-867-5; MDL No: MFCD00149186; RTECS: WE6660000
Crystalline; Linear Formula: Na2S2O3·5H2O; MW: 248.17
Melting Point: 48°
Density (g/mL): 1.69
- RTECS: WE6660000
- Crystalline
Formula:
Na₂S₂O₃.5H₂O Melting Pt: 48 °C Density: 1.69 g/mL |
MDL Number:
MFCD00149186 CAS Number: 10102-17-7 EINECS: 231-867-5 |
Specification Test Results
Assay | 99% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...