(R)-(-)-Methyl mandelate ≥99%

Supplier: BeanTown Chemical

Synonyms: D(-)-Methyl glycolate, (R)-(-)-Methyl glycolate, (R)-(-)-Mandelic acid methyl ester, D(-)-Methyl mandelate, D(-)-2-Hydroxy-2-phenylacetic acid methyl ester, D(-)-Mandelic acid methyl ester, (R)-Methyl 2-hydroxy-2-phenylacetate, Methyl (R)-(-)-mandelate, (R)-(-)-2-Hydroxy-2-phenylacetic acid methyl ester

143645-1G 143645-5G
BT143645-1GEA 32 USD
BT143645-1G BT143645-5G
(R)-(-)-Methyl mandelate ≥99%
(R)-(-)-Methyl mandelate

CAS: 20698-91-3; MDL No: MFCD00064247 Powder; Molecular Formula: C9H10O3 ; MW: 166.17 Melting Point: 56-58° Optical Rotation: [α]20/D -144°, c = 1 in methanol


  • Optical rotation: [α]20/D -144°, c = 1 in methanol
  • Powder

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
Boiling Pt: 267
Melting Pt: 56…58 °C
MDL Number: MFCD00064247
CAS Number: 20698-91-3

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR