Barium sulfate ≥97%

Supplier: BeanTown Chemical

143640-1KG 143640-5KG
BT143640-1KGEA 48.83 USD
BT143640-1KG BT143640-5KG
Barium sulfate ≥97%
Barium sulphate

CAS: 7727-43-7; EC No: 231-784-4; MDL No: MFCD00003455 1-4 Micron Powder; Linear Formula: BaSO4; MW: 233.39


  • 1-4 Micron Powder

Formula: BaSO₄
MW: 233.39 g/mol
Boiling Pt: 1600
Melting Pt: 1345
MDL Number: MFCD00003455
CAS Number: 7727-43-7
EINECS: 231-784-4
Merck Index: 13,00997

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR