D-(+)-Sucrose
Supplier: BeanTown Chemical
Synonyms:
alpha-D-Glucopyranoside, β-ᴅ-Fructofuranosyl-α-ᴅ-glucopyranoside, α-ᴅ-Glucopyranosyl β-ᴅ-fructofuranoside, D(+)-Saccharose, beta-D-Fructofuranosyl alpha-D-glucopyranoside, α-ᴅ-Glc-(1→2)-β-ᴅ-Fru, D-(+)-Saccharose, Sucrose, Sugar
143635-10KG
143635-2.5KG
143635-500G
BT143635-10KGEA
219
USD
BT143635-10KG
BT143635-2.5KG
BT143635-500G
D-(+)-Sucrose
D(+)-Sucrose
CAS: 57-50-1; EC No: 200-334-9; MDL No: MFCD00006626; RTECS: WN6500000
Crystalline; Molecular Formula: C12H22O11; MW: 342.30
Melting Point: 185-187°
Optical Rotation: [α]20/D +66.5±1°, c = 10 in water
- RTECS: WN6500000
- Optical rotation: [α]20/D +66.5±1°, c = 10 in water
- Crystalline
Formula:
C₁₂H₂₂O₁₁ Melting Pt: 185…187 °C |
MDL Number:
MFCD00006626 CAS Number: 57-50-1 EINECS: 200-334-9 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...