Benzylamine ≥98%

Supplier: BeanTown Chemical

Synonyms: α-Aminotoluene

143625-1KG 143625-500G
BT143625-1KGEA 107 USD
BT143625-1KG BT143625-500G
Benzylamine ≥98%
Benzylamine

CAS: 100-46-9; EC No: 202-854-1; MDL No: MFCD00008106; RTECS: DP1488500 UN No: UN2735; Haz Class: 8; Packing Group: II Liquid; Linear Formula: C6H5CH2NH2; Molecular Formula: C7H9N; MW: 107.15 Melting Point: 10°; Boiling Point: 184-185°; Flash point: 65°C (149°F) Density (g/mL): 0.981; Refractive Index: 1.543 Air Sensitive


  • RTECS: DP1488500
  • Refractive index: 1.543
  • Air Sensitive
  • Liquid

Formula: (C₆H₅)CH₂NH₂
MW: 107.16 g/mol
Boiling Pt: 185
Melting Pt: 10
Density: 0.981 g/mL
Flash Pt: 65 °C (149 °F)
MDL Number: MFCD00008106
CAS Number: 100-46-9
EINECS: 202-854-1
UN: 2735
ADR: 8 II
Merck Index: 13,01127

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR