Benzylamine ≥98%
Supplier: BeanTown Chemical
Synonyms:
α-Aminotoluene
143625-1KG
143625-500G
BT143625-1KGEA
107
USD
BT143625-1KG
BT143625-500G
Benzylamine ≥98%
Benzylamine
CAS: 100-46-9; EC No: 202-854-1; MDL No: MFCD00008106; RTECS: DP1488500
UN No: UN2735; Haz Class: 8; Packing Group: II
Liquid; Linear Formula: C6H5CH2NH2; Molecular Formula: C7H9N; MW: 107.15
Melting Point: 10°; Boiling Point: 184-185°; Flash point: 65°C (149°F)
Density (g/mL): 0.981; Refractive Index: 1.543
Air Sensitive
- RTECS: DP1488500
- Refractive index: 1.543
- Air Sensitive
- Liquid
Formula:
(C₆H₅)CH₂NH₂ Boiling Pt: 184…185 °C Melting Pt: 10 °C Density: 0.981 g/mL Flash Pt: 65 °C (149 °F) |
MDL Number:
MFCD00008106 CAS Number: 100-46-9 EINECS: 202-854-1 UN: 2735 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...