Tin(IV) fluoride ≥99%
Supplier: BeanTown Chemical
143505-1G
143505-5G
BT143505-1GEA
41.55
USD
BT143505-1G
BT143505-5G
Tin(IV) fluoride ≥99%
Tin (IV) fluoride
CAS: 7783-62-2; EC No: 232-016-0; MDL No: MFCD00042541
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II
-6 Mesh Crystalline; Linear Formula: SnF4; MW: 194.70
Melting Point: 705° (sublimes)
Moisture Sensitive
- Moisture Sensitive
- -6 Mesh Crystalline
Formula:
SnF₄ Melting Pt: 705 °C (subl.) |
MDL Number:
MFCD00042541 CAS Number: 7783-62-2 EINECS: 232-016-0 ADR: 8 (6.1),II |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...