4-Hydroxybenzoic acid ≥98%
Supplier: BeanTown Chemical
143445-1KG
143445-250G
143445-5KG
BT143445-1KGEA
65
USD
BT143445-1KG
BT143445-250G
BT143445-5KG
4-Hydroxybenzoic acid ≥98%
4-Hydroxybenzoic acid
CAS: 99-96-7; EC No: 202-804-9; MDL No: MFCD00002547; RTECS: DH1925000
Solid; Molecular Formula: C7H6O3; MW: 138.12
Melting Point: 213-217°
- RTECS: DH1925000
- Solid
Formula:
C₇H₆O₃ Melting Pt: 213…217 °C |
MDL Number:
MFCD00002547 CAS Number: 99-96-7 EINECS: 202-804-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...