4-Methyl-2-pentanone ≥99.0% for HPLC
Supplier: BeanTown Chemical
Synonyms:
Methyl iso-Butyl Ketone, Isobutyl Methyl Ketone, Methyl Isobutyl Ketone, MIK
143420-1L
143420-4L
143420-4X1L
143420-4X4L
BT143420-1LEA
73.8
USD
BT143420-1L
BT143420-4L
BT143420-4X1L
BT143420-4X4L
4-Methyl-2-pentanone ≥99.0% for HPLC
4-Methyl-2-pentanone
CAS: 108-10-1; EC No: 203-550-1; MDL No: MFCD00008938; RTECS: SA9275000
UN No: UN1245; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6H12O; MW: 100.16
Melting Point: -80°; Boiling Point: 117-118°; Flash point: 14°C (57°F)
Density (g/mL): 0.801; Refractive Index: 1.395
- RTECS: SA9275000
- Refractive index: 1.395
- Liquid
Formula:
(CH₃)₂CHCH₂COCH₃ Boiling Pt: 117…118 °C Melting Pt: –80 °C Density: 0.801 g/mL Flash Pt: 14 °C (57 °F) |
MDL Number:
MFCD00008938 CAS Number: 108-10-1 EINECS: 203-550-1 UN: 1245 ADR: 3,II |
|
Specification Test Results
Assay | 99.0% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...