Butyramide ≥98%
Supplier: BeanTown Chemical
143370-100G
143370-500G
BT143370-100GEA
65
USD
BT143370-100G
BT143370-500G
Butyramide ≥98%
Butyramide
CAS: 541-35-5; EC No: 208-776-4; MDL No: MFCD00041894
Powder/Flakes; Linear Formula: CH3CH2CH2CONH2; Molecular Formula: C4H9NO; MW: 87.12
Melting Point: 114-116°
- Powder/Flakes
Formula:
C₄H₉NO Melting Pt: 114…116 °C |
MDL Number:
MFCD00041894 CAS Number: 541-35-5 EINECS: 208-776-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...