1-Cyclohexene-1-acetic acid

Supplier: BeanTown Chemical

Synonyms: 1-Cyclohexenylacetic acid

143330-1G 143330-25G 143330-5G
BT143330-1GEA 30 USD
BT143330-1G BT143330-25G BT143330-5G
1-Cyclohexene-1-acetic acid
1-Cyclohexene-1-acetic acid

CAS: 18294-87-6; EC No: 242-172-1; MDL No: MFCD00015482 Liquid/Low Melting Solid; Molecular Formula: C8H12O2; MW: 140.18 Melting Point: 32-35° Density (g/mL): 1.04


  • Liquid/Low Melting Solid

Formula: C₈H₁₂O₂
Melting Pt: 32…35 °C
Density: 1.04 g/mL
MDL Number: MFCD00015482
CAS Number: 18294-87-6
EINECS: 242-172-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR