2-Adamantanol ≥98%
Supplier: BeanTown Chemical
Synonyms:
2-Hydroxytricyclo[3.3.1.1(3,7)]decane, Tricyclo[3.3.1.1(3,7)]decan-2-ol, 2-Tricyclo[3.3.1.1(3,7)]decanol
143320-25G
143320-5G
BT143320-25GEA
219
USD
BT143320-25G
BT143320-5G
2-Adamantanol ≥98%
2-Adamantanol
CAS: 700-57-2; EC No: 211-846-7; MDL No: MFCD00074744
Crystalline/Powder; Molecular Formula: C10H16O; MW: 152.23
Melting Point: 258-262°
- Crystalline/Powder
Formula:
C₁₀H₁₆O Melting Pt: 258…262 °C |
MDL Number:
MFCD00074744 CAS Number: 700-57-2 EINECS: 211-846-7 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...