Ethanol denatured, anhydrous 94.0-96.0% ACS

Supplier: BeanTown Chemical
Danger

Synonyms: Ethyl alcohol denatured

143215-1L 143215-4L 143215-500ML
BT143215-1LEA 56.93 USD
BT143215-1L BT143215-4L BT143215-500ML
Ethanol denatured, anhydrous 94.0-96.0% ACS
Ethanol denatured

CAS: 64-17-5; EC No: 200-578-6; MDL No: MFCD00003568; RTECS: KQ6300000 UN No: UN1987; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CH2OH; Molecular Formula: C2H6O; MW: 46.08 Melting Point: -90°; Boiling Point: 78°; Flash point: 11°C (52°F) Density (g/mL): 0.789 Hygroscopic


  • RTECS: KQ6300000
  • Hygroscopic

Formula: H₃CCH₂OH
MW: 46.07 g/mol
Boiling Pt: 78 °C
Melting Pt: –90 °C
Density: 0.789 g/mL
Flash Pt: 11 °C
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3 II

Order Now

Specification Test Results

Assay 94.0-96.0%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR