Copper ≥99.5% (trace metals basis), spherical powder -100 mesh

Supplier: BeanTown Chemical

143185-1KG 143185-250G 143185-50G
BT143185-1KGEA 98 USD
BT143185-1KG BT143185-250G BT143185-50G
Copper ≥99.5% (trace metals basis), spherical powder -100 mesh
Copper

CAS: 7440-50-8; EC No: 231-159-6; MDL No: MFCD00010965 UN No: UN3089; Haz Class: 4.1; Packing Group: III Powder; Molecular Formula: Cu; MW: 63.55 Melting Point: 1083.4°; Boiling Point: 2567° Density (g/mL): 8.94


  • Powder

Formula: Cu
Boiling Pt: 2567 °C
Melting Pt: 1083.4 °C
Density: 8.94 g/mL
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
UN: 3089
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR