N,N-Dimethylglycine ethyl ester ≥98%
Supplier: BeanTown Chemical
Synonyms:
Di-Me-Gly-OEt
143165-25G
BT143165-25GEA
63
USD
BT143165-25G
N,N-Dimethylglycine ethyl ester ≥98%
N,N-Dimethylglycine ethyl ester
CAS: 33229-89-9; EC No: 251-411-9; MDL No: MFCD00009175; RTECS: MB9990000
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: (CH3)2NCH2COOCH2CH3; Molecular Formula: C6H13NO2; MW: 131.17
Boiling Point: 150-151°; Flash point: 32°C (90°F)
Density (g/mL): 0.928; Refractive Index: 1.413
Air Sensitive, Hygroscopic, Moisture Sensitive
- RTECS: MB9990000
- Refractive index: 1.413
- Air Sensitive, Hygroscopic, Moisture Sensitive
- Liquid
Formula:
C₆H₁₃NO₂ MW: 131.17 g/mol Boiling Pt: 150…151 °C Density: 0.928 g/mL Flash Pt: 32 °C (90 °F) |
MDL Number:
MFCD00009175 CAS Number: 33229-89-9 EINECS: 251-411-9 ADR: 3 III |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...