5-Aminolevulinic acid hydrochloride ≥99%
Supplier: BeanTown Chemical
Synonyms:
5-Amino-4-ketovaleric acid hydrochloride, 5-Amino-4-oxopentanoic acid hydrochloride, δ-Aminolevulinic Acid Hydrochloride
143115-100MG
143115-2G
143115-500MG
BT143115-100MGEA
32
USD
BT143115-100MG
BT143115-2G
BT143115-500MG
5-Aminolevulinic acid hydrochloride ≥99%
5-Aminolevulinic acid hydrochloride
CAS: 5451-09-2; EC No: 226-679-5; MDL No: MFCD00012869; RTECS: OI1640000
Crystalline; Molecular Formula: C5H9NO3·HCl; MW: 167.59
Melting Point: 150° (decomposes)
Hygroscopic
- RTECS: OI1640000
- Hygroscopic
- Crystalline
Formula:
C₅H₁₀ClNO₃ Melting Pt: 150 °C (dec.) |
MDL Number:
MFCD00012869 CAS Number: 5451-09-2 EINECS: 226-679-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...