Indole-3-carboxylic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
1H-Indole-3-carboxylic acid
143060-1G
143060-25G
143060-5G
BT143060-1GEA
33
USD
BT143060-1G
BT143060-25G
BT143060-5G
Indole-3-carboxylic acid ≥98%
Indole-3-carboxylic acid
CAS: 771-50-6; EC No: 212-231-6; MDL No: MFCD00005624; RTECS: NK7882892
Powder; Molecular Formula: C9H6NO2; MW: 161.16
Melting Point: 232-234° (decomposes)
- RTECS: NK7882892
- Powder
Formula:
C₉H₇NO₂ Melting Pt: 232…234 °C (dec.) |
MDL Number:
MFCD00005624 CAS Number: 771-50-6 EINECS: 212-231-6 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...