5-Chloro-2-nitropyridine ≥97%

Supplier: BeanTown Chemical

143050-1G 143050-25G 143050-5G
BT143050-1GEA 57 USD
BT143050-1G BT143050-25G BT143050-5G
5-Chloro-2-nitropyridine ≥97%
5-Chloro-2-nitropyridine

CAS: 52092-47-4; MDL No: MFCD03092916 Solid; Molecular Formula: C5H3ClN2O2 ; MW: 158.54 Melting Point: 119-123°


  • Solid

Formula: C₅H₃ClN₂O₂
Melting Pt: 119…123 °C
MDL Number: MFCD03092916
CAS Number: 52092-47-4

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR