Sulfuric acid ≥92%

Supplier: BeanTown Chemical

143035-100ML 143035-500ML
BT143035-100MLEA 140 USD
BT143035-100ML BT143035-500ML
Sulfuric acid ≥92%
Sulfuric acid

CAS: 7664-93-9; EC No: 231-639-5; MDL No: MFCD00064589; RTECS: WS5600000 UN No: UN1830; Haz Class: 8; Packing Group: II Liquid; Linear Formula: H2SO4; MW: 98.08 Melting Point: 3°; Boiling Point: 290° Density (g/mL): 1.84


  • RTECS: WS5600000
  • Liquid

Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84 g/mL
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II

Order Now

Specification Test Results

Assay 99.9999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR