2,4-Dichlorobenzoyl chloride ≥98%
Supplier: BeanTown Chemical
142995-250G
142995-50G
BT142995-250GEA
112
USD
BT142995-250G
BT142995-50G
2,4-Dichlorobenzoyl chloride ≥98%
2,4-Dichlorobenzoyl chloride
CAS: 89-75-8; EC No: 201-936-4; MDL No: MFCD00000661; RTECS: DM6636766
UN No: UN3265; Haz Class: 8; Packing Group: II
Liquid; Molecular Formula: C7H3Cl3O ; MW: 209.46
Melting Point: 16-18°; Boiling Point: 150°/34 mmHg; Flash point: 137°C (238°F)
Density (g/mL): 1.494; Refractive Index: 1.592
Moisture Sensitive
- RTECS: DM6636766
- Refractive index: 1.592
- Moisture Sensitive
- Liquid
Formula:
C₇H₃Cl₃O Boiling Pt: 150 °C/34 mmHg Melting Pt: 16…18 °C Density: 1.494 g/mL Flash Pt: 137 °C (238 °F) |
MDL Number:
MFCD00000661 CAS Number: 89-75-8 EINECS: 201-936-4 UN: 3265 ADR: 8,II |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...