n-Tetracosane ≥99%
Supplier: BeanTown Chemical
Synonyms:
N-tetracosane
142955-100G
142955-25G
142955-5G
BT142955-100GEA
108.57
USD
BT142955-100G
BT142955-25G
BT142955-5G
n-Tetracosane ≥99%
n-Tetracosane
CAS: 646-31-1; EC No: 211-474-5; MDL No: MFCD00009352
Crystalline/Powder; Linear Formula: CH3(CH2)22CH3; Molecular Formula: C24H50; MW: 338.66
Melting Point: 49-52°; Boiling Point: 391°; Flash point:
- Crystalline/Powder
Formula:
H₃C(CH₂)₂₂CH₃ MW: 338.66 g/mol Boiling Pt: 391.3 Melting Pt: 49…52 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00009352 CAS Number: 646-31-1 EINECS: 211-474-5 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...