2'-Fluoro-4'-methoxyacetophenone ≥99%
Supplier: BeanTown Chemical
142930-100G
142930-25G
142930-5G
BT142930-100GEA
660
USD
BT142930-100G
BT142930-25G
BT142930-5G
2'-Fluoro-4'-methoxyacetophenone ≥99%
2'-Fluoro-4'-methoxyacetophenone
CAS: 74457-86-6; MDL No: MFCD00042290
Solid; Molecular Formula: C9H9FO2; MW: 168.16
Melting Point: 52-54°; Flash point:
- Solid
Formula:
C₉H₉FO₂ Melting Pt: 52…54 °C Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00042290 CAS Number: 74457-86-6 |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...