1-Bromo-3-chloro-5-fluorobenzene ≥98%
Supplier: BeanTown Chemical
Synonyms:
3-Chloro-5-fluorobromobenzene
142900-100G
142900-25G
142900-5G
BT142900-100GEA
1035
USD
BT142900-100G
BT142900-25G
BT142900-5G
1-Bromo-3-chloro-5-fluorobenzene ≥98%
1-Bromo-3-chloro-5-fluorobenzene
CAS: 33863-76-2; MDL No: MFCD00070747
Liquid; Molecular Formula: C6H3BrClF; MW: 209.45
Density (g/mL): 1.72
- Liquid
Formula:
C₆H₃BrClF Density: 1.72 g/mL |
MDL Number:
MFCD00070747 CAS Number: 33863-76-2 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...