Indole ≥99%

Supplier: BeanTown Chemical

Synonyms: 1H-Indole

142845-1KG 142845-250G 142845-50G
BT142845-1KGEA 429 USD
BT142845-1KG BT142845-250G BT142845-50G
Indole ≥99%
Indole

CAS: 120-72-9; EC No: 204-420-7; MDL No: MFCD00005607; RTECS: NL2450000 UN No: UN2811; Haz Class: 6.1; Packing Group: III Solid; Molecular Formula: C8H7N; MW: 117.15 Melting Point: 51-54°; Boiling Point: 253-254°; Flash point: 121°C (249°F) Light Sensitive


  • RTECS: NL2450000
  • Light Sensitive
  • Solid

Formula: C₈H₇N
MW: 117.15 g/mol
Boiling Pt: 253…254 °C
Melting Pt: 51…54 °C
Flash Pt: 121 °C (249 °F)
MDL Number: MFCD00005607
CAS Number: 120-72-9
EINECS: 204-420-7
UN: 2811
ADR: 6.1 III
Merck Index: 13,04985

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR