Tristearin (glycerol tristearate) ≥99%

Supplier: BeanTown Chemical

Synonyms: Propane-1,2,3-triyl tristearate, Tristearin

142805-25G 142805-5G
BT142805-25GEA 687 USD
BT142805-25G BT142805-5G
Tristearin (glycerol tristearate) ≥99%
Tristearin (Glycerol tristearate)

CAS: 555-43-1; EC No: 209-097-6; MDL No: MFCD00036230 Powder; Linear Formula: [CH3(CH2)16COOCH2]2CHOCO(CH2)16CH3; Molecular Formula: C57H110O6; MW: 891.48 Melting Point: 72°; Boiling Point: 260°


  • Powder

Formula: C₅₇H₁₁₀O₆
Boiling Pt: 260 °C
Melting Pt: 72 °C
MDL Number: MFCD00036230
CAS Number: 555-43-1
EINECS: 209-097-6

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR