Zinc sulfide ≥99.995%
Supplier: BeanTown Chemical
Synonyms:
Zinc (II) sulphide, Zinc (II) sulfide
142800-10G
142800-250G
142800-50G
BT142800-10GEA
25
USD
BT142800-10G
BT142800-250G
BT142800-50G
Zinc sulfide ≥99.995%
Zinc sulphide
CAS: 1314-98-3; EC No: 215-251-3; MDL No: MFCD00011301; RTECS: ZH5400000
3-12mm Pieces; Linear Formula: ZnS; MW: 97.43
Melting Point: 1700°
Density (g/mL): 4.1
- RTECS: ZH5400000
- 3-12mm Pieces
Formula:
SZn MW: 97.46 g/mol Melting Pt: 1064 Density: 4.1 g/mL |
MDL Number:
MFCD00011301 CAS Number: 1314-98-3 EINECS: 215-251-3 Merck Index: 12,10294 |
Specification Test Results
Assay | 99.995% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...