L(+)-Tartaric acid ≥99%
Supplier: BeanTown Chemical
Synonyms:
L-Tartaric Acid, (2R,3R)-(+)-Tartaric acid, Tartaric Acid, (R,R)-(+)-2,3-Dihydroxysuccinic acid, (R,R)-(+)-Tartaric acid, (2R,3R)-2,3-Dihydroxysuccinic acid
142795-100G
142795-2.5KG
142795-500G
BT142795-100GEA
54.47
USD
BT142795-100G
BT142795-2.5KG
BT142795-500G
L(+)-Tartaric acid ≥99%
L(+)-Tartaric acid
CAS: 87-69-4; EC No: 201-766-0; MDL No: MFCD00064207; RTECS: WW7875000
Crystalline; Molecular Formula: C4H6O6; MW: 150.09
Melting Point: 170-172°
Optical Rotation: [α]20/D +13.5±0.5°, c = 10% in water
- RTECS: WW7875000
- Optical rotation: [α]20/D +13.5±0.5°, c = 10% in water
- Crystalline
Formula:
C₄H₆O₆ MW: 150.09 g/mol Boiling Pt: 399 Melting Pt: 170…172 °C Flash Pt: 150 |
MDL Number:
MFCD00064207 CAS Number: 87-69-4 EINECS: 201-766-0 Merck Index: 13,09158 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...