Silicon carbide ≥99% (trace metals basis)
Supplier: BeanTown Chemical
142710-2KG
142710-500G
BT142710-2KGEA
482
USD
BT142710-2KG
BT142710-500G
Silicon carbide ≥99% (trace metals basis)
Silicon carbide
CAS: 409-21-2; EC No: 206-991-8; MDL No: MFCD00049531; RTECS: VW0450000
-325 Mesh Powder; Linear Formula: SiC; MW: 40.10
Melting Point: 2700°
Density (g/mL): 3.22
- RTECS: VW0450000
- -325 Mesh Powder
Formula:
SiC Melting Pt: 2700 °C Density: 3.22 g/mL |
MDL Number:
MFCD00049531 CAS Number: 409-21-2 EINECS: 206-991-8 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...