Zinc(II) dibutyldithiocarbamate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Dibutyldithiocarbamic Acid Zinc(II) Salt
142660-25G
BT142660-25GEA
37.55
USD
BT142660-25G
Zinc(II) dibutyldithiocarbamate ≥97%
Zinc(II) dibutyldithiocarbamate
CAS: 136-23-2; EC No: 205-232-8; MDL No: MFCD00067274; RTECS: ZH0175000
Solid; Molecular Formula: C18H36N2S4Zn; MW: 474.14
Melting Point: 104-110°
- RTECS: ZH0175000
- Solid
Formula:
C₉H₁₉NS₂ Melting Pt: 104…110 °C |
MDL Number:
MFCD00067274 CAS Number: 136-23-2 EINECS: 205-232-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...