Ammonium hydrogen sulfate ≥99.9%

Supplier: BeanTown Chemical

Synonyms: Ammonium bisulfate, Ammonium bisulphate

142650-100G 142650-25G 142650-500G
BT142650-100GEA 76 USD
BT142650-100G BT142650-25G BT142650-500G
Ammonium hydrogen sulfate ≥99.9%
Ammonium hydrogen sulphate

CAS: 7803-63-6; EC No: 232-265-5; MDL No: MFCD00051073 UN No: UN2506; Haz Class: 8; Packing Group: II Lump; Linear Formula: NH4HSO4; MW: 115.11 Melting Point: 121-145°; Boiling Point: 350° (decomposes) Density (g/mL): 1.79 Hygroscopic


  • Hygroscopic
  • Lump

Formula: H₅NO₄S
MW: 115.11 g/mol
Boiling Pt: 350 (dec.)
Melting Pt: 138.85
Density: 1.79 g/mL
MDL Number: MFCD00051073
CAS Number: 7803-63-6
EINECS: 232-265-5
UN: 2506
ADR: 8 II
Merck Index: 12,00526

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR