Glycine ≥98.5% ACS

Supplier: BeanTown Chemical

Synonyms: Aminoacetic acid, Glycocoll, H-Gly-OH

142580-100G 142580-2.5KG 142580-500G
BT142580-100GEA 45.92 USD
BT142580-100G BT142580-2.5KG BT142580-500G
Glycine ≥98.5% ACS
Glycine

CAS: 56-40-6; EC No: 200-272-2; MDL No: MFCD00008131; RTECS: MB7600000 Crystalline ; Linear Formula: NH2CH2CO2H; MW: 75.07 Melting Point: 240° (decomposes)


  • RTECS: MB7600000
  • Crystalline

Formula: NH₂CH₂COOH
Melting Pt: 240 °C (dec.)
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d944007d30d5d4650062014a1b152bd9bfb66ea3&vid=093f4227cbdd1c4c6515ef9c4b85d93799877dcc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.5% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR