Glycine ≥98.5% ACS
Supplier: BeanTown Chemical
Synonyms:
Aminoacetic acid, Glycocoll, H-Gly-OH
142580-100G
142580-2.5KG
142580-500G
BT142580-100GEA
45.92
USD
BT142580-100G
BT142580-2.5KG
BT142580-500G
Glycine ≥98.5% ACS
Glycine
CAS: 56-40-6; EC No: 200-272-2; MDL No: MFCD00008131; RTECS: MB7600000
Crystalline ; Linear Formula: NH2CH2CO2H; MW: 75.07
Melting Point: 240° (decomposes)
- RTECS: MB7600000
- Crystalline
Formula:
NH₂CH₂COOH Melting Pt: 240 °C (dec.) |
MDL Number:
MFCD00008131 CAS Number: 56-40-6 EINECS: 200-272-2 |
|
Specification Test Results
Assay | 98.5% min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...