Ethyl-2-phenylacrylate ≥94%

Supplier: BeanTown Chemical

Synonyms: Ethyl 2-phenylacrylate

142515-1G 142515-25G 142515-5G
BT142515-1GEA 88 USD
BT142515-1G BT142515-25G BT142515-5G
Ethyl-2-phenylacrylate ≥94%
Ethyl-2-phenylacrylate

CAS: 22286-82-4; EC No: 244-892-1; MDL No: MFCD00026862 Liquid; Molecular Formula: C11H12O2; MW: 176.22 Boiling Point: 123-125°/16 mmHg Light Sensitive


  • Light Sensitive
  • Liquid

Formula: C₁₁H₁₂O₂
MW: 176.215 g/mol
Boiling Pt: 123…125 °C/16 mmHg
MDL Number: MFCD00026862
CAS Number: 22286-82-4
EINECS: 244-892-1

Order Now

Specification Test Results

Assay 94%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR