Hexaboron silicide ≥98%

Supplier: BeanTown Chemical

Synonyms: Boron silicide

142295-10G 142295-50G
BT142295-10GEA 95 USD
BT142295-10G BT142295-50G
Hexaboron silicide ≥98%
Hexaboron silicide

CAS: 12008-29-6; EC No: 234-535-8; MDL No: MFCD00180907 -200 Mesh Powder; Linear Formula: SiB6; MW: 92.95


  • -200 Mesh Powder

Formula: B₆Si
MW: 92.95 g/mol
MDL Number: MFCD00180907
CAS Number: 12008-29-6
EINECS: 234-535-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR