Dexamethasone ≥98%
Supplier: BeanTown Chemical
142255-1G
142255-25G
142255-5G
BT142255-1GEA
127.1
USD
BT142255-1G
BT142255-25G
BT142255-5G
Dexamethasone ≥98%
Dexamethasone
CAS: 50-02-2; EC No: 200-003-9; MDL No: MFCD00064136; RTECS: TU3980000
Powder; Molecular Formula: C22H29FO5; MW: 392.46
Melting Point: 262-264°
Light Sensitive
- RTECS: TU3980000
- Light Sensitive
- Powder
Formula:
C₂₂H₂₉FO₅ Melting Pt: 262…264 °C |
MDL Number:
MFCD00064136 CAS Number: 50-02-2 EINECS: 200-003-9 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...